accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 28240-66-6

HPHP

CAS RN 28240-66-6

IUPAC Name: phenyl[3-(phenylphosphanyl)propyl]phosphane
Synonyms: phenyl[3-(phenylphosphanyl)propyl]phosphane
Molecular Weight (g/mol): 260.26
Molecular Formula: C15H18P2
InChi Key: AVNRJUHUOZDFKS-UHFFFAOYSA-N
SMILES: C(CPC1=CC=CC=C1)CPC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)