accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOONH

CAS RN 28226-22-4

IUPAC Name: 6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 6-nitro-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: GZAJZBARYACGSO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2OCCNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results