accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONONCl

CAS RN 28163-00-0

IUPAC Name: 2-chloro-4-nitrobenzonitrile
Synonyms: 2-chloro-4-nitrobenzonitrile
Molecular Weight (g/mol): 182.56
Molecular Formula: C7H3ClN2O2
InChi Key: ZIGQFRQZYVQQDR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C#N)C(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)