CAS RN 2809353-52-2
CAS RN 2809353-52-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ LL-K9-3
Tocris Bioscience™ LL-K9-3
Selective HyT-based degrader of the CDK9-cyclin T1 complex