accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 280749-17-9

CH3H3CSONNOH3CNOO(E/Z)

CAS RN 280749-17-9

IUPAC Name: 1-(4-acetylpiperazin-1-yl)-3-(3-nitro-4-{[2-(propan-2-yl)phenyl]sulfanyl}phenyl)prop-2-en-1-one
Synonyms: 1-(4-acetylpiperazin-1-yl)-3-{4-[(2-isopropylphenyl)sulfanyl]-3-nitrophenyl}prop-2-en-1-one
Molecular Weight (g/mol): 453.56
Molecular Formula: C24H27N3O4S
InChi Key: HTGGAYLWTDOFDK-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1SC1=C(C=C(C=CC(=O)N2CCN(CC2)C(C)=O)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results