CAS RN 280560-78-3
CAS RN 280560-78-3
IUPAC Name:
1-[(4-bromophenyl)methyl]-4-methyl-1,4-diazepane
Synonyms:
1-[(4-bromophenyl)methyl]-4-methyl-1,4-diazepane
Molecular Weight (g/mol):
283.21
Molecular Formula:
C13H19BrN2
InChi Key:
WQUWRVLHCKJAMG-UHFFFAOYSA-N
SMILES:
CN1CCCN(CC2=CC=C(Br)C=C2)CC1
1
–
1
of
1
results
1
1-(4-Bromobenzyl)-4-methylperhydro-1,4-diazepine, 97%, Thermo Scientific™
CAS: 280560-78-3 Molecular Formula: C13H19BrN2 Molecular Weight (g/mol): 283.213 MDL Number: MFCD00810550 InChI Key: WQUWRVLHCKJAMG-UHFFFAOYSA-N Synonym: 1-4-bromobenzyl-4-methylperhydro-1,4-diazepine,1-4-bromophenyl methyl-4-methyl-1,4-diazepane,1-4-bromobenzyl-4-methyl-1,4-diazepane,cambridge id 5258520,1h-1,4-diazepine,1-4-bromophenyl methyl hexahydro-4-methyl PubChem CID: 763047 IUPAC Name: 1-[(4-bromophenyl)methyl]-4-methyl-1,4-diazepane SMILES: CN1CCCN(CC1)CC2=CC=C(C=C2)Br