CAS RN 27973-29-1
CAS RN 27973-29-1
IUPAC Name:
1,6-dibromopyrene
Synonyms:
1,6-dibromopyrene
Molecular Weight (g/mol):
360.05
Molecular Formula:
C16H8Br2
InChi Key:
JRCJYPMNBNNCFE-UHFFFAOYSA-N
SMILES:
BrC1=CC=C2C=CC3=C(Br)C=CC4=CC=C1C2=C34