accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OO(S)NH2HNOOHCl

CAS RN 27894-50-4

IUPAC Name: methyl 2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoate hydrochloridyl
Synonyms: methyl 2-amino-6-{[(benzyloxy)carbonyl]amino}hexanoate hydrochloridyl
Molecular Weight (g/mol): 329.8
Molecular Formula: C15H22ClN2O4
InChi Key: RUIMMNNJOYMHFC-UHFFFAOYNA-N
SMILES: [Cl].COC(=O)C(N)CCCCNC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results