accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SnOOOOH3CCH3H3CH3CCH3H3C

CAS RN 2781-10-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 521.37
Molecular Formula: C24H50O4Sn
InChi Key: POLVRWSQZMVOIP-UHFFFAOYNA-N
SMILES: CCCC[SnH2]CCCC.CCCCC(CC)C(O)=O.CCCCC(CC)C(O)=O
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)