accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 27774-13-6

OSOOOOV

CAS RN 27774-13-6

IUPAC Name: oxovanadiumbis(ylium) sulfate
Synonyms: vanadyl sulfate vanadin(IV) oxide sulfate oxo[sulfato(2-)-O]-vanadium vanadium oxide sulphate vanadium oxysulfate vanadic sulfate
Molecular Weight (g/mol): 163
Molecular Formula: O5SV
InChi Key: UUUGYDOQQLOJQA-UHFFFAOYSA-L
SMILES: O=[V++].[O-]S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results