CAS RN 2776191-07-0
CAS RN 2776191-07-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ BromoTag™ AGB3
Tocris Bioscience™ BromoTag™ AGB3
Selective Degrader of Brd4BD2 L387A fusion proteins (BromoTag™)