accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOCH

CAS RN 2774-84-7

IUPAC Name: undec-10-yn-1-ol
Synonyms: undec-10-yn-1-ol
Molecular Weight (g/mol): 168.28
Molecular Formula: C11H20O
InChi Key: YUQZOUNRPZBQJK-UHFFFAOYSA-N
SMILES: OCCCCCCCCCC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)