accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClSOOHNOO

CAS RN 27685-90-1

IUPAC Name: 2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride
Synonyms: 2-oxo-3H-1,3-benzoxazole-6-sulfonyl chloride
Molecular Weight (g/mol): 233.62
Molecular Formula: C7H4ClNO4S
InChi Key: IBHCNMJSVDRBSY-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C2NC(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results