CAS RN 2768-02-7
CAS RN 2768-02-7
IUPAC Name:
ethenyltrimethoxysilane
Synonyms:
silane, ethenyltrimethoxy-
Molecular Weight (g/mol):
148.23
Molecular Formula:
C5H12O3Si
InChi Key:
NKSJNEHGWDZZQF-UHFFFAOYSA-N
SMILES:
CO[Si](OC)(OC)C=C
1
–
3
of
3
results
1
Vinyltrimethoxysilane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Vinyltrimethoxysilane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Vinyltrimethoxysilane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.