accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2766-43-0

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]-3-hydroxypropanoate
Molecular Weight (g/mol): 219.24
Molecular Formula: C9H17NO5
InChi Key: SANNKFASHWONFD-UHFFFAOYNA-N
SMILES: COC(=O)C(CO)NC(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results