accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OSOON

CAS RN 27655-40-9

IUPAC Name: isoquinoline-5-sulfonate
Synonyms: isoquinoline-5-sulfonate
Molecular Weight (g/mol): 208.21
Molecular Formula: C9H6NO3S
InChi Key: YFMJTLUPSMCTOQ-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C1=C2C=CN=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results