accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2765-11-9

H3CO

CAS RN 2765-11-9

IUPAC Name: pentadecanal
Synonyms: pentadecanal
Molecular Weight (g/mol): 226.4
Molecular Formula: C15H30O
InChi Key: XGQJZNCFDLXSIJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results