CAS RN 2764704-18-7
CAS RN 2764704-18-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ LSN 3318839
Tocris Bioscience™ LSN 3318839
Glucagon-like peptide-1 receptor (GLP1-R) positive allosteric modulator