accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2743-40-0

H3C(S)NH2OOCH3CH3HCl

CAS RN 2743-40-0

IUPAC Name: hydrogen ethyl 2-amino-4-methylpentanoate chloride
Synonyms: hydrogen leucine, ethyl ester chloride
Molecular Weight (g/mol): 195.69
Molecular Formula: C8H18ClNO2
InChi Key: NOUDPBCEONUCOV-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C(N)CC(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)