accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 274-09-9

IUPAC Name: 2H-1,3-benzodioxole
Synonyms: benzodioxole methylenedioxybenzene 1,3-dioxindan o-(methylenedioxy)benzene
Molecular Weight (g/mol): 122.12
Molecular Formula: C7H6O2
InChi Key: FTNJQNQLEGKTGD-UHFFFAOYSA-N
SMILES: C1OC2=CC=CC=C2O1
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)