CAS RN 2732614-50-3
CAS RN 2732614-50-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Remifentanil Ester Hydrochloride, TRC
CAS: 2732614-50-3 Molecular Formula: C13H18N2O2 • HCl Molecular Weight (g/mol): 270.76 Synonym: (Phenylamino)methyl Piperidine-4-carboxylate,Methyl 4-anilinopiperidine-4-carboxylate Hydrochloride IUPAC Name: methyl 4-(phenylamino)piperidine-4-carboxylate hydrochloride SMILES: COC(C1(NC2=CC=CC=C2)CCNCC1)=O.Cl