accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

SCNN

CAS RN 2719-32-6

IUPAC Name: 4-isothiocyanatobenzonitrile
Synonyms: 4-isothiocyanatobenzonitrile
Molecular Weight (g/mol): 160.19
Molecular Formula: C8H4N2S
InChi Key: DZFKAXLNKZXNHD-UHFFFAOYSA-N
SMILES: S=C=NC1=CC=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results