accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONH

CAS RN 2703-17-5

IUPAC Name: methyl 1H-pyrrole-3-carboxylate
Synonyms: methyl 1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: WLBNVSIQCFHAQB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CNC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results