accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 270082-22-9

ClHOO(S)HNH

CAS RN 270082-22-9

IUPAC Name: hydrogen 2-[(3S)-1,2,3,4-tetrahydroisoquinolin-3-yl]acetic acid chloride
Synonyms: hydrogen (3S)-1,2,3,4-tetrahydroisoquinolin-3-ylacetic acid chloride
Molecular Weight (g/mol): 227.69
Molecular Formula: C11H14ClNO2
InChi Key: SWYPWAIQEURSFY-PPHPATTJSA-N
SMILES: [H+].[Cl-].OC(=O)C[C@@H]1CC2=CC=CC=C2CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results