accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 26970-82-1

H2OH2OH2OH2OH2ONaNaOSeOO

CAS RN 26970-82-1

IUPAC Name: disodium pentahydrate selenite
Synonyms: disodium pentahydrate selenite
Molecular Weight (g/mol): 263.02
Molecular Formula: H10Na2O8Se
InChi Key: TUANAMBRHOLYTH-UHFFFAOYSA-L
SMILES: O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results