accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H2NH2N

CAS RN 2687-25-4

IUPAC Name: 3-methylbenzene-1,2-diamine
Synonyms: 2,3-diaminotoluene
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: AXNUJYHFQHQZBE-UHFFFAOYSA-N
SMILES: CC1=CC=CC(N)=C1N
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)