accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2683-43-4

NH2ClClNOO

CAS RN 2683-43-4

IUPAC Name: 2,4-dichloro-6-nitroaniline
Synonyms: 2,4-dichloro-6-nitroaniline
Molecular Weight (g/mol): 207.01
Molecular Formula: C6H4Cl2N2O2
InChi Key: IZEZAMILKKYOPW-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(Cl)C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)