accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONHNHN

CAS RN 26725-51-9

IUPAC Name: 2,4-dihydro-1H-1,2,3-benzotriazol-4-one
Synonyms: 1,2-dihydro-1,2,3-benzotriazol-4-one
Molecular Weight (g/mol): 135.13
Molecular Formula: C6H5N3O
InChi Key: NPZTUJOABDZTLV-UHFFFAOYSA-N
SMILES: O=C1C=CC=C2NNN=C12
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)