accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2651-55-0

IUPAC Name: 3-ethoxy-4-methoxybenzoate
Synonyms: 3-ethoxy-4-methoxybenzoate
Molecular Weight (g/mol): 195.2
Molecular Formula: C10H11O4
InChi Key: DMSAIFTWQMXOBE-UHFFFAOYSA-M
SMILES: CCOC1=CC(=CC=C1OC)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results