accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOHOH

CAS RN 26446-35-5

IUPAC Name: 2,3-dihydroxypropyl acetate
Synonyms: acetin acetin, 1-mono-
Molecular Weight (g/mol): 134.13
Molecular Formula: C5H10O4
InChi Key: KMZHZAAOEWVPSE-UHFFFAOYNA-N
SMILES: CC(=O)OCC(O)CO
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results