CAS RN 2642-63-9
IUPAC Name:
1-(3,4-dichlorophenyl)ethan-1-one
Synonyms:
1-(3,4-dichlorophenyl)ethanone
Molecular Weight (g/mol):
189.04
Molecular Formula:
C8H6Cl2O
InChi Key:
WBPAOUHWPONFEQ-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(Cl)C(Cl)=C1
1
–
2
of
2
results
1
3',4'-Dichloroacetophenone, 98+%
CAS: 2642-63-9 Molecular Formula: C8H6Cl2O Molecular Weight (g/mol): 189.04 MDL Number: MFCD00000553 InChI Key: WBPAOUHWPONFEQ-UHFFFAOYSA-N Synonym: 3',4'-dichloroacetophenone,1-3,4-dichlorophenyl ethanone,3,4-dichloroacetophenone,1-3,4-dichlorophenyl ethan-1-one,ethanone, 1-3,4-dichlorophenyl,acetophenone, 3',4'-dichloro,1-acetyl-3,4-dichlorobenzene,3,4 dichloro acetophenone,pubchem3383,3,4-dicloroacetophenone PubChem CID: 75841 IUPAC Name: 1-(3,4-dichlorophenyl)ethanone SMILES: CC(=O)C1=CC=C(Cl)C(Cl)=C1