accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CSOOOO

CAS RN 26272-83-3

IUPAC Name: oxetan-3-yl 4-methylbenzene-1-sulfonate
Synonyms: oxetan-3-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 228.26
Molecular Formula: C10H12O4S
InChi Key: UMFWNFVHKAJOSE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC1COC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results