accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 262587-05-3

FFOBr

CAS RN 262587-05-3

IUPAC Name: 1-bromo-3-(difluoromethoxy)benzene
Synonyms: 1-bromo-3-(difluoromethoxy)benzene
Molecular Weight (g/mol): 223.02
Molecular Formula: C7H5BrF2O
InChi Key: NWBDGKNKAFGQQL-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results