accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HBrNH2

CAS RN 26227-54-3

IUPAC Name: hydrogen cyclohexanamine bromide
Synonyms: hydrogen cyclohexylamine bromide
Molecular Weight (g/mol): 180.09
Molecular Formula: C6H14BrN
InChi Key: QOHWJRRXQPGIQW-UHFFFAOYSA-N
SMILES: [H+].[Br-].NC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results