accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 261951-81-9

FFFFN

CAS RN 261951-81-9

IUPAC Name: 3-fluoro-2-(trifluoromethyl)benzonitrile
Synonyms: 3-fluoro-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 189.11
Molecular Formula: C8H3F4N
InChi Key: ZLFIVXFFYDWCNG-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C#N)=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results