accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 261951-75-1

CH3FOOH

CAS RN 261951-75-1

IUPAC Name: 2-(5-fluoro-2-methylphenyl)acetic acid
Synonyms: (5-fluoro-2-methylphenyl)acetic acid
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: BPWHUOQJJZVQLO-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results