accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 261951-68-2

CH3NH2F

CAS RN 261951-68-2

IUPAC Name: 1-(4-fluoro-3-methylphenyl)methanamine
Synonyms: 1-(4-fluoro-3-methylphenyl)methanamine
Molecular Weight (g/mol): 139.17
Molecular Formula: C8H10FN
InChi Key: PQXWYQZQCWWMDQ-UHFFFAOYSA-N
SMILES: CC1=CC(CN)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results