accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH

CAS RN 26186-02-7

IUPAC Name: tridec-1-yne
Synonyms: tridec-1-yne
Molecular Weight (g/mol): 180.34
Molecular Formula: C13H24
InChi Key: GZEDKDBFUBPZNG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)