accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH2OFF

CAS RN 261763-35-3

IUPAC Name: 2,3-difluoro-4-methylbenzamide
Synonyms: 2,3-difluoro-4-methylbenzamide
Molecular Weight (g/mol): 171.15
Molecular Formula: C8H7F2NO
InChi Key: JWPKXGWPXBRCLL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(N)=O)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results