accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 261762-59-8

HOFCl

CAS RN 261762-59-8

IUPAC Name: (2-chloro-5-fluorophenyl)methanol
Synonyms: (2-chloro-5-fluorophenyl)methanol
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: VTZKFYRHXVQMQL-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results