accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2612-26-2

H3COHOH

CAS RN 2612-26-2

IUPAC Name: 2-butylpropane-1,3-diol
Synonyms: 2-butylpropane-1,3-diol
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: SIMYRXPCIUXDGR-UHFFFAOYSA-N
SMILES: CCCCC(CO)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results