accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 26107-80-2

CH3OOFFFFFF

CAS RN 26107-80-2

IUPAC Name: methyl 3,5-bis(trifluoromethyl)benzoate
Synonyms: methyl 3,5-bis(trifluoromethyl)benzoate
Molecular Weight (g/mol): 272.15
Molecular Formula: C10H6F6O2
InChi Key: LTYNBDAJUXZFHP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results