accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OONHOO

CAS RN 26054-60-4

IUPAC Name: benzyl N-(2-oxooxetan-3-yl)carbamate
Synonyms: benzyl N-(2-oxooxetan-3-yl)carbamate
Molecular Weight (g/mol): 221.21
Molecular Formula: C11H11NO4
InChi Key: CWFZPRQDHIUBDO-UHFFFAOYNA-N
SMILES: O=C(NC1COC1=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results