accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClONHOO

CAS RN 26049-94-5

IUPAC Name: benzyl N-(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamate
Synonyms: benzyl N-(4-chloro-3-oxo-1-phenylbutan-2-yl)carbamate
Molecular Weight (g/mol): 331.8
Molecular Formula: C18H18ClNO3
InChi Key: OYHLRJGDELITAF-UHFFFAOYNA-N
SMILES: ClCC(=O)C(CC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results