accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 2596-47-6

IUPAC Name: (2E)-3-[4-(acetyloxy)-3-methoxyphenyl]prop-2-enoic acid
Synonyms: (2E)-3-[4-(acetyloxy)-3-methoxyphenyl]prop-2-enoic acid
Molecular Weight (g/mol): 236.22
Molecular Formula: C12H12O5
InChi Key: IHKNVZISLLDMOR-GQCTYLIASA-N
SMILES: COC1=CC(\C=C\C(O)=O)=CC=C1OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results