accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHSOOHO

CAS RN 2580-77-0

IUPAC Name: 2-(2-hydroxyethanesulfonyl)ethan-1-ol
Synonyms: 2-(2-hydroxyethanesulfonyl)ethanol
Molecular Weight (g/mol): 154.18
Molecular Formula: C4H10O4S
InChi Key: QQLILYBIARWEIF-UHFFFAOYSA-N
SMILES: OCCS(=O)(=O)CCO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results