accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 25764-10-7

NLa

CAS RN 25764-10-7

IUPAC Name: lanthanioamine
Synonyms: lanthanioamine
Molecular Weight (g/mol): 152.91
Molecular Formula: LaN
InChi Key: QCLQZCOGUCNIOC-UHFFFAOYSA-N
SMILES: N#[La]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results