accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFOOOFFF

CAS RN 25753-15-5

IUPAC Name: 3-(trifluoromethyl)benzoyl 3-(trifluoromethyl)benzoate
Synonyms: 3-(trifluoromethyl)benzoyl 3-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 362.23
Molecular Formula: C16H8F6O3
InChi Key: BYLMYFXAIMTZIC-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results