accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CO

CAS RN 2571-39-3

IUPAC Name: (3,4-dimethylphenyl)(phenyl)methanone
Synonyms: (3,4-dimethylphenyl)(phenyl)methanone
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: JENOLWCGNVWTJN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1C)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results