accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OH3CH3CH3COHOH3C

CAS RN 256934-39-1

IUPAC Name: {4-[2,6-dimethoxy-4-(2-methyloctan-2-yl)phenyl]-6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl}methanol
Synonyms: {4-[2,6-dimethoxy-4-(2-methyloctan-2-yl)phenyl]-6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl}methanol
Molecular Weight (g/mol): 414.63
Molecular Formula: C27H42O3
InChi Key: CFMRIVODIXTERW-UHFFFAOYNA-N
SMILES: CCCCCCC(C)(C)C1=CC(OC)=C(C2C=C(CO)C3CC2C3(C)C)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results